ADVH49B82324 AChemBlock

tert-butyl 1,7-diazaspiro[3.5]nonane-7-carboxylate hemioxalate

  • Cas Number : 1523617-82-4
  • Molecular Weight : 542.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-1-7-diazaspiro-3-5-nonane-7-carboxylate-hemioxalate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79564
Availability details for
Quantity
  • Availability
  • Add to Cart