ADVH49B82325 AChemBlock

3,3-difluoro-1-methylcyclobutan-1-amine hydrochloride

  • Cas Number : 1523606-39-4
  • Molecular Weight : 157.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-3-difluoro-1-methylcyclobutan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79565
Availability details for
Quantity
  • Availability
  • Add to Cart