ADVH49B82327 AChemBlock

3-aminocyclobutane-1-carbonitrile hydrochloride

  • Cas Number : 1523572-04-4
  • Molecular Weight : 132.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-aminocyclobutane-1-carbonitrile-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79567
Availability details for
Quantity
  • Availability
  • Add to Cart