ADVH49B82328 AChemBlock

tert-butyl n-[(3-methylazetidin-3-yl)methyl]carbamate hemioxalate

  • Cas Number : 1523572-03-3
  • Molecular Weight : 490.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-3-methylazetidin-3-yl-methyl-carbamate-hemioxalate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79568
Availability details for
Quantity
  • Availability
  • Add to Cart