ADVH49B82329 AChemBlock

2,8-dioxa-5-azaspiro[3.5]nonane hemioxalate

  • Cas Number : 1523571-80-3
  • Molecular Weight : 348.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-8-dioxa-5-azaspiro-3-5-nonane-hemioxalate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79570
Availability details for
Quantity
  • Availability
  • Add to Cart