ADVH49B8232B AChemBlock

methyl (2R,4R)-2-methylpiperidine-4-carboxylate hydrochloride

  • Cas Number : 1523530-24-6
  • Molecular Weight : 193.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-2r-4r-2-methylpiperidine-4-carboxylate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79573
Availability details for
Quantity
  • Availability
  • Add to Cart