ADVH49B8232E AChemBlock

tert-butyl 4-(5-amino-1H-indol-3-yl)piperidine-1-carboxylate

  • Cas Number : 151273-40-4
  • Molecular Weight : 315.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-5-amino-1h-indol-3-yl-piperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79613
Availability details for
Quantity
  • Availability
  • Add to Cart