ADVH49B82333 AChemBlock

8-(chloromethoxy)-5-nitro-quinoline

  • Cas Number : 1501350-97-5
  • Molecular Weight : 238.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/8-chloromethoxy-5-nitro-quinoline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79669
Availability details for
Quantity
  • Availability
  • Add to Cart