ADVH49B82337 AChemBlock

6-chloro-N-methylpyridazin-3-amine

  • Cas Number : 14959-32-1
  • Molecular Weight : 143.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-n-methylpyridazin-3-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71672
Availability details for
Quantity
  • Availability
  • Add to Cart