ADVH49B82338 AChemBlock

methyl 7,7-difluoronorcarane-3-carboxylate

  • Cas Number : 1494594-75-0
  • Molecular Weight : 190.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-7-7-difluoronorcarane-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79695
Availability details for
Quantity
  • Availability
  • Add to Cart