ADVH49B8233D AChemBlock

tert-butyl 3-(2,4-difluorophenyl)-3-hydroxyazetidine-1-carboxylate

  • Cas Number : 1482460-15-0
  • Molecular Weight : 285.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-2-4-difluorophenyl-3-hydroxyazetidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79733
Availability details for
Quantity
  • Availability
  • Add to Cart