ADVH49B8233E AChemBlock

4,7-dibromo-1H-1,3-benzodiazole

  • Cas Number : 148185-66-4
  • Molecular Weight : 275.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-7-dibromo-1h-1-3-benzodiazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79736
Availability details for
Quantity
  • Availability
  • Add to Cart