ADVH49B82343 AChemBlock

3-methyl-7-(trifluoromethyl)-3,4-dihydro-1H-quinoxalin-2-one

  • Cas Number : 146741-08-4
  • Molecular Weight : 230.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-methyl-7-trifluoromethyl-3-4-dihydro-1h-quinoxalin-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79775
Availability details for
Quantity
  • Availability
  • Add to Cart