ADVH49B82346 AChemBlock

2-Bromomethyl-6-pyridine carboxylic acidmethyl ester

  • Cas Number : 146462-25-1
  • Molecular Weight : 230.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-bromomethyl-6-pyridine-carboxylic-acidmethyl-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86509
Availability details for
Quantity
  • Availability
  • Add to Cart