ADVH49B82347 AChemBlock

3-Prop-2-ynyl-azetidine-1-carboxylic acid tert-butyl ester

  • Cas Number : 1463502-41-1
  • Molecular Weight : 195.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-prop-2-ynyl-azetidine-1-carboxylic-acid-tert-butyl-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86642
Availability details for
Quantity
  • Availability
  • Add to Cart