ADVH49B8234B AChemBlock

5-bromo-4-oxo-pentanoic acid

  • Cas Number : 14594-23-1
  • Molecular Weight : 195.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-4-oxo-pentanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79795
Availability details for
Quantity
  • Availability
  • Add to Cart