ADVH49B8234C AChemBlock

SH-4-54

  • Cas Number : 1456632-40-8
  • Molecular Weight : 610.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/sh-4-54.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72378
Availability details for
Quantity
  • Availability
  • Add to Cart