ADVH49B8234F AChemBlock

methyl 2-[(1R)-1-[(2-aminopyridin-3-yl)oxy]ethyl]-4-fluorobenzoate

  • Cas Number : 1454847-99-4
  • Molecular Weight : 290.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-1r-1-2-aminopyridin-3-yl-oxy-ethyl-4-fluorobenzoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79810
Availability details for
Quantity
  • Availability
  • Add to Cart