ADVH49B82350 AChemBlock

4-methylthiazole-2-carboxylic acid

  • Cas Number : 14542-16-6
  • Molecular Weight : 143.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-methylthiazole-2-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71683
Availability details for
Quantity
  • Availability
  • Add to Cart