ADVH49B82352 AChemBlock

(5S)-5-phenylmorpholin-2-one

  • Cas Number : 144896-92-4
  • Molecular Weight : 177.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5s-5-phenylmorpholin-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79835
Availability details for
Quantity
  • Availability
  • Add to Cart