ADVH49B82356 AChemBlock

6-bromo-1-chloro-5-fluoroisoquinoline

  • Cas Number : 1445564-11-3
  • Molecular Weight : 260.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-1-chloro-5-fluoroisoquinoline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79868
Availability details for
Quantity
  • Availability
  • Add to Cart