ADVH49B82359 AChemBlock

(3S,4R)-3-fluoropiperidin-4-ol hydrochloride

  • Cas Number : 1443380-89-9
  • Molecular Weight : 155.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3s-4r-3-fluoropiperidin-4-ol-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79893
Availability details for
Quantity
  • Availability
  • Add to Cart