ADVH49B8235A AChemBlock

6-chloro-1-isopropyl-pyrazolo[3,4-d]pyrimidine

  • Cas Number : 1443289-75-5
  • Molecular Weight : 196.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-1-isopropyl-pyrazolo-3-4-d-pyrimidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79895
Availability details for
Quantity
  • Availability
  • Add to Cart