ADVH49B8235B AChemBlock

5-(1-Naphthyl)cyclohexane-1,3-dione

  • Cas Number : 144128-68-7
  • Molecular Weight : 238.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-1-naphthyl-cyclohexane-1-3-dione.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86775
Availability details for
Quantity
  • Availability
  • Add to Cart