ADVH49B8235E AChemBlock

ethyl 5-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate

  • Cas Number : 1436686-17-7
  • Molecular Weight : 270.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-5-bromopyrazolo-1-5-a-pyrimidine-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79933
Availability details for
Quantity
  • Availability
  • Add to Cart