ADVH49B8235F AChemBlock

(1R,2R)-ethyl 2-aminocyclohexanecarboxylate hydrochloride

  • Cas Number : 1436-60-8
  • Molecular Weight : 207.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1r-2r-ethyl-2-aminocyclohexanecarboxylate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72441
Availability details for
Quantity
  • Availability
  • Add to Cart