ADVH49B82360 AChemBlock

methyl 5-amino-2,4-dihydroxy-benzoate

  • Cas Number : 1435323-67-3
  • Molecular Weight : 183.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-amino-2-4-dihydroxy-benzoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79936
Availability details for
Quantity
  • Availability
  • Add to Cart