ADVH49B82362 AChemBlock

tert-butyl (3S)-3-methyl-4-(6-nitropyridin-3-yl)piperazine-1-carboxylate

  • Cas Number : 1433849-53-6
  • Molecular Weight : 322.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3s-3-methyl-4-6-nitropyridin-3-yl-piperazine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79949
Availability details for
Quantity
  • Availability
  • Add to Cart