ADVH49B82366 AChemBlock

2-methoxy-3,5-dimethyl-pyridin-4-amine

  • Cas Number : 1429510-61-1
  • Molecular Weight : 152.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-methoxy-3-5-dimethyl-pyridin-4-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79986
Availability details for
Quantity
  • Availability
  • Add to Cart