ADVH49B82368 AChemBlock

(5-bromo-2-methoxypyridin-3-yl)methanamine hydrochloride

  • Cas Number : 1428532-91-5
  • Molecular Weight : 253.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-2-methoxypyridin-3-yl-methanamine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71688
Availability details for
Quantity
  • Availability
  • Add to Cart