ADVH49B82369 AChemBlock

Methyl 4-fluoro-1H-indazole-3-carboxylate

  • Cas Number : 1427504-03-7
  • Molecular Weight : 194.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-fluoro-1h-indazole-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72425
Availability details for
Quantity
  • Availability
  • Add to Cart