ADVH49B8236B AChemBlock

3-(difluoromethyl)piperidine hydrochloride

  • Cas Number : 1427455-16-0
  • Molecular Weight : 171.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-difluoromethyl-piperidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80015
Availability details for
Quantity
  • Availability
  • Add to Cart