ADVH49B82370 AChemBlock

2-(1-tert-butoxycarbonyl-3-methyl-azetidin-3-yl)acetic acid

  • Cas Number : 1422344-49-7
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-1-tert-butoxycarbonyl-3-methyl-azetidin-3-yl-acetic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80079
Availability details for
Quantity
  • Availability
  • Add to Cart