ADVH49B82372 AChemBlock

6-bromo-8-iodo-2-methyl-imidazo[1,2-a]pyridine

  • Cas Number : 1421601-69-5
  • Molecular Weight : 336.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-8-iodo-2-methyl-imidazo-1-2-a-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80085
Availability details for
Quantity
  • Availability
  • Add to Cart