ADVH49B82373 AChemBlock

N1-(2-(dimethylamino)ethyl)-5-methoxy-N1-methyl-N4-(4-(1-methyl-1H-indol-3-yl)pyrimidin-2-yl)benzene-1,2,4-triamine

  • Cas Number : 1421372-66-8
  • Molecular Weight : 445.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/n1-2-dimethylamino-ethyl-5-methoxy-n1-methyl-n4-4-1-methyl-1h-indol-3-yl-pyrimidin-2-yl-benzene-1-2-4-triamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86469
Availability details for
Quantity
  • Availability
  • Add to Cart