ADVH49B82374 AChemBlock

1,2,3,4-tetrahydropyrido[1,2-a]pyrazin-6-one;hydrochloride

  • Cas Number : 1421065-64-6
  • Molecular Weight : 186.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-2-3-4-tetrahydropyrido-1-2-a-pyrazin-6-one-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80093
Availability details for
Quantity
  • Availability
  • Add to Cart