ADVH49B82375 AChemBlock

N-(1-acetyl-3,3-dimethylindolin-6-yl)-2-chloronicotinamide

  • Cas Number : 1420999-91-2
  • Molecular Weight : 343.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-1-acetyl-3-3-dimethylindolin-6-yl-2-chloronicotinamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86698
Availability details for
Quantity
  • Availability
  • Add to Cart