ADVH49B82376 AChemBlock

1-(3-bromo-4-(hydroxymethyl)phenyl)ethanone

  • Cas Number : 1418144-62-3
  • Molecular Weight : 229.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-3-bromo-4-hydroxymethyl-phenyl-ethanone.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86220
Availability details for
Quantity
  • Availability
  • Add to Cart