ADVH49B8237B AChemBlock

(2-fluoro-4-phenoxyphenyl)boronic acid

  • Cas Number : 1414356-30-1
  • Molecular Weight : 232.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-fluoro-4-phenoxyphenyl-boronic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80149
Availability details for
Quantity
  • Availability
  • Add to Cart