ADVH49B8237C AChemBlock

(1S,2R)-2-fluorocyclopropan-1-amine hydrochloride

  • Cas Number : 141042-20-8
  • Molecular Weight : 111.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1s-2r-2-fluorocyclopropan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80159
Availability details for
Quantity
  • Availability
  • Add to Cart