ADVH49B8237E AChemBlock

tert-butyl N-(azepan-4-ylmethyl)carbamate

  • Cas Number : 1408074-78-1
  • Molecular Weight : 228.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-azepan-4-ylmethyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80169
Availability details for
Quantity
  • Availability
  • Add to Cart