ADVH49B8237F AChemBlock

tert-butyl 2,8-diazaspiro[4.5]decane-8-carboxylate oxalate

  • Cas Number : 1408074-53-2
  • Molecular Weight : 330.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-8-diazaspiro-4-5-decane-8-carboxylate-oxalate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71696
Availability details for
Quantity
  • Availability
  • Add to Cart