ADVH49B82381 AChemBlock

7-bromo-2,3-dihydro-1-benzofuran-3-ol

  • Cas Number : 1404230-46-1
  • Molecular Weight : 215.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-bromo-2-3-dihydro-1-benzofuran-3-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80180
Availability details for
Quantity
  • Availability
  • Add to Cart