ADVH49B82383 AChemBlock

ethyl 8-oxo-5,6,7,9-tetrahydropyrrolo[1,2-a]azepine-2-carboxylate

  • Cas Number : 1400927-61-8
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-8-oxo-5-6-7-9-tetrahydropyrrolo-1-2-a-azepine-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80201
Availability details for
Quantity
  • Availability
  • Add to Cart