ADVH49B82384 AChemBlock

tert-butyl (1R,4R)-5-oxo-2-azabicyclo[2.2.1]heptane-2-carboxylate

  • Cas Number : 1400808-00-5
  • Molecular Weight : 211.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-1r-4r-5-oxo-2-azabicyclo-2-2-1-heptane-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80203
Availability details for
Quantity
  • Availability
  • Add to Cart