ADVH49B82387 AChemBlock

2-benzyl-2,6-diazaspiro[3.3]heptane; oxalic acid

  • Cas Number : 1394840-24-4
  • Molecular Weight : 278.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-benzyl-2-6-diazaspiro-3-3-heptane-oxalic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80231
Availability details for
Quantity
  • Availability
  • Add to Cart