ADVH49B82388 AChemBlock

3-Ethynyl-3-hydroxy-1-methyl-piperidin-2-one

  • Cas Number : 1394119-72-2
  • Molecular Weight : 153.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-ethynyl-3-hydroxy-1-methyl-piperidin-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86631
Availability details for
Quantity
  • Availability
  • Add to Cart