ADVH49B8238A AChemBlock

6-(pyridin-2-yl)imidazo[2,1-b][1,3]thiazole-5-carbaldehyde

  • Cas Number : 139359-78-7
  • Molecular Weight : 229.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-pyridin-2-yl-imidazo-2-1-b-1-3-thiazole-5-carbaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80255
Availability details for
Quantity
  • Availability
  • Add to Cart