ADVH49B8238C AChemBlock

(1S,3S,5S)-rel-2-tert-butoxycarbonyl-5-methyl-2-azabicyclo[3.1.0]hexane-3-carboxylic acid

  • Cas Number : 1393537-79-5
  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1s-3s-5s-rel-2-tert-butoxycarbonyl-5-methyl-2-azabicyclo-3-1-0-hexane-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80276
Availability details for
Quantity
  • Availability
  • Add to Cart