ADVH49B82390 AChemBlock

5H,6H,7H,8H-imidazo[1,2-a]pyridine-6-carboxylic acid hydrochloride

  • Cas Number : 139183-99-6
  • Molecular Weight : 202.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5h-6h-7h-8h-imidazo-1-2-a-pyridine-6-carboxylic-acid-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80293
Availability details for
Quantity
  • Availability
  • Add to Cart